EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Evaluation of comprehensive two-dimensional gas chromatography with accurate mass time-of-flight mass spectrometry for the metabolic profiling of plant-fungus interaction in Aquilaria malaccensis.
PMID 25704770