PloS one
A rapid and convenient method for fluorescence analysis of in vitro cultivated metacestode vesicles from Echinococcus multilocularis.
PMID 25705880
Abstract
We here describe a convenient method for preparation, fixation and fluorescence analysis of in vitro cultivated metacestode vesicles from E. multilocularis. Parasite materials could be prepared in one hour, did not need to be sectioned, and were subsequently utilized for further whole-mount staining assays directly. Using these preparations, in combination with conventional fluorescence staining techniques, we could detect the expression and subcellular localization of a specific protein and identify in situ proliferative or apoptotic cells in the germinal layer of metacestode vesicles. Based on this approach, future molecular and cellular analysis of Echinococcus metacestode vesicles in the in vitro system will be greatly facilitated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11684809910
In Situ Cell Death Detection Kit, AP