EMAIL THIS PAGE TO A FRIEND
Journal of Pineal Research
Melatonin-mediated Bim up-regulation and cyclooxygenase-2 (COX-2) down-regulation enhances tunicamycin-induced apoptosis in MDA-MB-231 cells.
PMID 25711465