EMAIL THIS PAGE TO A FRIEND
Investigative Ophthalmology & Visual Science
Poly(ADP-Ribose) Polymerase Inhibition Improves Corneal Epithelial Innervation and Wound Healing in Diabetic Rats.
PMID 25711635