EMAIL THIS PAGE TO A FRIEND
Activitas nervosa superior
Oxyprothepin and clorotepin compared in the hyperkinetic syndrome.
PMID 2571227
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O111
Octoclothepin maleate salt, solid
C19H21ClN2S · C4H4O4