EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Cardiomyocyte lipotoxicity is mediated by Il-6 and causes down-regulation of PPARs.
PMID 25712520
Abstract
Here we sought to evaluate the effect of palmitate on cytokine and PPAR activity/expression. We investigated the effect of BSA conjugated palmitate and oleate on PPAR activity, PPAR-α and δ expression, as well as the expression of cytokines and key factors responsible for β-oxidation by qRT-PCR and western blotting in primary rat neonatal cardiomyocytes (NCMs). Furthermore we evaluated the effect of anti-inflammatory actions of AICAR and PPAR agonists on cytokine expression and cell death in palmitate treated NCMs. We found that palmitate caused down regulation of PPARs and increased cytokine expression and cell death, all of which was significantly attenuated by the co-administration of either AICAR or PPAR agonists. This work supports the pro-inflammatory actions of intracellular lipid and provides further insight into the pathological mechanism of cardiac lipotoxicity as occurs in diabetic hearts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O7501
Sodium oleate, ≥99%
C18H33NaO2
O3880
Sodium oleate, ≥95% (capillary GC)
C18H33NaO2