EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Plasma metabolite profiling and chemometric analyses of lung cancer along with three controls through gas chromatography-mass spectrometry.
PMID 25712604