EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Mono(boratabenzene) rare-earth metal dialkyl complexes: synthesis, structure and catalytic behaviors for styrene polymerization.
PMID 25714489
Abstract
Four mono(boratabenzene) rare-earth metal dialkyl complexes, [(3,5-Me2-C5H3BR)Ln(CH2SiMe3)2(THF)] (1: R = NEt2, Ln = Sc; 2: R = NEt2, Ln = Lu; 3: R = Ph, Ln = Sc; 4: R = Ph, Ln = Lu), were synthesized efficiently via a one-pot strategy with Li[3,5-Me2-C5H3BR] (R = NEt2, Ph), LnCl3(THF)x (Ln = Sc, x = 3; Ln = Lu, x = 0), and LiCH2SiMe3. The solid-state structures of 1 and 2 were determined by single-crystal X-ray diffraction. Variable-temperature NMR studies indicated that the energy barrier for the rotation of aminoboratabenzene in 1 (ΔG‡ ≈ 71 kJ mol−1) is higher than that of phenylboratabenzene in 3 (ΔG‡ ≈ 59 kJ mol−1). These mono(boratabenzene) rare-earth metal dialkyl complexes’ catalytic behaviors for styrene polymerization were investigated, and found that mono(boratabenzene) scandium dialkyl complexes show high catalytic activities for syndiotactic polymerization upon activation with cocatalysts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
101346
Dichlorophenylborane, 97%
C6H5BCl2