EMAIL THIS PAGE TO A FRIEND
Journal of medical virology
Chloroquine enhances replication of influenza A virus A/WSN/33 (H1N1) in dose-, time-, and MOI-dependent manners in human lung epithelial cells A549.
PMID 25715935
Abstract
Anti-malaria drug, chloroquine, has been reported to be effective against influenza A virus (IAV) in vitro and used in in-vivo experiments and clinical trial for prevention or treatment of influenza. In this study, it has been shown by immunofluorescence, hemagglutination, and plaque assays that chloroquine enhanced A/WSN/33 (H1N1) replication with pronounced cytopathic effect in dose-, time-, and MOI-dependent manners in human lung epithelial cells A549. Time-of-addition assay showed that inhibitory effect on virus replication by chloroquine pre-treatment was indistinctive, and virus productions were enhanced when the drug was applied after viral adsorption. The effectiveness of chloroquine as an anti-influenza drug is questioned, and caution in its use is recommended.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6628
Chloroquine diphosphate salt, solid, ≥98%
C18H26ClN3 · 2H3PO4