EMAIL THIS PAGE TO A FRIEND
Journal of Materials Science. Materials in Medicine
Chitosan-based electrospun nanofibrous mats, hydrogels and cast films: novel anti-bacterial wound dressing matrices.
PMID 25716023