EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
Aging alters long-term potentiation--related gene networks and impairs synaptic protein synthesis in the rat hippocampus.
PMID 25716081
Abstract
During aging, memory retention and persistence of long-term potentiation (LTP) are impaired, suggesting an aging-related deterioration in mechanisms regulating information storage. Late-phase LTP requires synthesis of proteins at synapses as well as integrated regulation of gene networks. Because aging diminishes the persistence of LTP, primarily by affecting the transition between early and late phases, we assessed whether this was reflected in perturbation of gene networks. Using DNA microarray analysis, we compared LTP-associated gene expression in young (5xa0months), middle-aged (15xa0months), and old (22xa0months) male Sprague-Dawley rats. As expected, we found no significant difference in LTP measured 20xa0minutes postinduction; however, we found that overall more genes were regulated in the young group. Bioinformatics predicted not only dysregulation of activator protein-1 and nuclear factor kB transcription factor activity and epigenetic modifications but also dysregulation of protein synthesis. Notably, we confirmed an age-related impairment in metabotropic and ionotropic receptor-mediated synaptic protein synthesis. Together, these results demonstrate that LTP-specific gene expression is altered with aging and suggest that dysregulation of synaptic protein synthesis also contributes to the age-dependent reduction in LTP persistence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0378
Chloramphenicol, ≥98% (HPLC)
C11H12Cl2N2O5
C7795
Chloramphenicol, γ-irradiated
C11H12Cl2N2O5
C1919
Chloramphenicol, BioReagent, suitable for plant cell culture
C11H12Cl2N2O5
C0857
Chloramphenicol, meets USP testing specifications
C11H12Cl2N2O5
C1863
Chloramphenicol, Biotechnology Performance Certified, suitable for plant cell culture
C11H12Cl2N2O5
561932
Deblock trichloroacetic acid solution, 3 % (w/v) in methylene chloride
C2HCl3O2
M9500
DL-Methionine, ≥99%
C5H11NO2S
M2768
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
C5H11NO2S
W330108
DL-Methionine, 99%, FCC, FG
C5H11NO2S
672920
TCA Deblock, 3 % (w/v) in methylene chloride
C2HCl3O2
T9159
Trichloroacetic acid, BioXtra, ≥99.0%
C2HCl3O2
T6399
Trichloroacetic acid, ACS reagent, ≥99.0%
C2HCl3O2
T4885
Trichloroacetic acid, ≥99.0% (titration)
C2HCl3O2
T8657
Trichloroacetic acid, for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
C2HCl3O2
91228
Trichloroacetic acid, BioUltra, ≥99.5% (T)
C2HCl3O2
91230
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
C2HCl3O2
27242
Trichloroacetic acid, puriss., meets analytical specification of Ph. Eur., USP 21, 99-100.5% (calc. to the dried substance)
C2HCl3O2
116114
Trichloroacetic acid, reagent grade, 98%
C2HCl3O2
T0324
Trichloroacetic acid solution, ~6.25 % (w/v), 0.38 N
C2HCl3O2
T0699
Trichloroacetic acid solution, 6.1 N
C2HCl3O2