EMAIL THIS PAGE TO A FRIEND
Journal of Immunology
Buprenorphine decreases the CCL2-mediated chemotactic response of monocytes.
PMID 25716997
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1570428
2-Propanol, United States Pharmacopeia (USP) Reference Standard
C3H8O
PHR1072
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O
91237
2-Propanol, analytical standard
C3H8O
90872
Acetone, analytical standard
C3H6O
1006801
Acetone, United States Pharmacopeia (USP) Reference Standard
C3H6O
PHR1219
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
C3H6O
48358
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
C3H6O
B0126
Bromophenol Blue
C19H10Br4O5S
114391
Bromophenol Blue, ACS reagent
C19H10Br4O5S
C6352
CTAP, ≥95%
C51H69N13O11S2
E7384
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
C26H35N5O6
49897
Formate standard for IC, 1.000 g/L in H2O, analytical standard
CH2O2
06473
Formic acid solution, BioUltra, 1.0 M in H2O
CH2O2
1295607
Glycerin, United States Pharmacopeia (USP) Reference Standard
C3H8O3
PHR1020
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O3
49779
Glycerol, tested according to Ph.Eur., anhydrous
C3H8O3
77067
Glycerol, analytical standard
C3H8O3
12146
Glycerol 3 M solution, 3 M
C3H8O3 · xH2O
I1149
Iodoacetamide, BioUltra
C2H4INO
I6125
Iodoacetamide, ≥99% (NMR), crystalline
C2H4INO
I5161
Iodoacetamide, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production
C2H4INO
71726
Sodium dodecyl sulfate, for ion pair chromatography, ≥99.0%
C12H25NaO4S
71727
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
C12H25NaO4S
Y0000620
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
C12H25NaO4S
74564
Trifluoroacetic acid, analytical standard
C2HF3O2
U0600000
Urea, European Pharmacopoeia (EP) Reference Standard
CH4N2O
1706698
Urea, United States Pharmacopeia (USP) Reference Standard
CH4N2O
56180
Urea, analytical standard
CH4N2O
51465
Urea, tested according to Ph.Eur.
CH4N2O
08582
Urea solution, for microbiology, 40% in H2O
CH4N2O