EMAIL THIS PAGE TO A FRIEND
The Analyst
Rapid detection of sugar alcohol precursors and corresponding nitrate ester explosives using direct analysis in real time mass spectrometry.
PMID 25717497
Abstract
This work highlights the rapid detection of nitrate ester explosives and their sugar alcohol precursors by direct analysis in real time mass spectrometry (DART-MS) using an off-axis geometry. Demonstration of the effect of various parameters, such as ion polarity and in-source collision induced dissociation (CID) on the detection of these compounds is presented. Sensitivity of sugar alcohols and nitrate ester explosives was found to be greatest in negative ion mode with sensitivities ranging from hundreds of picograms to hundreds of nanograms, depending on the characteristics of the particular molecule. Altering the in-source CID potential allowed for acquisition of characteristic molecular ion spectra as well as fragmentation spectra. Additional studies were completed to identify the role of different experimental parameters on the sensitivity for these compounds. Variables that were examined included the DART gas stream temperature, the presence of a related compound (i.e., the effect of a precursor on the detection of a nitrate ester explosive), incorporation of dopant species and the role of the analysis surface. It was determined that each variable affected the response and detection of both sugar alcohols and the corresponding nitrate ester explosives. From this work, a rapid and sensitive method for the detection of individual sugar alcohols and corresponding nitrate ester explosives, or mixtures of the two, has been developed, providing a useful tool in the real-world identification of homemade explosives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63560
D-Mannitol, ACS reagent, for microbiology, ≥99.0%
C6H14O6
17311
D-Mannitol, tested according to Ph Eur
C6H14O6
78513
D-Mannitol, ≥99.9999% (metals basis), for boron determination
C6H14O6
S1876
D-Sorbitol, ≥98%
C6H14O6
85529
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
C6H14O6
S6021
D-Sorbitol, for molecular biology, ≥98%
C6H14O6
S3889
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
C6H14O6
97336
D-Sorbitol, liquid, tested according to Ph Eur
C6H14O6
47841
D-Sorbitol, analytical standard
C6H14O6
S7547
D-Sorbitol, BioXtra, ≥98%
C6H14O6
W302902
D-Sorbitol, FCC, FG
C6H14O6
Y0000177
Erythritol, European Pharmacopoeia (EP) Reference Standard
C4H10O4
PHR1046
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O2
1265515
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
C2H6O2
85978
Ethylene glycol, analytical standard
C2H6O2
61941 Ethylene glycol 5 M solution
C2H6O2
721972 Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
C2H6O2
721980 Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
C2H6O2
PHR1351
Inositol, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6
PHR1007
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14O6
1375105
Mannitol, United States Pharmacopeia (USP) Reference Standard
C6H14O6
M0200000
Mannitol, European Pharmacopoeia (EP) Reference Standard
C6H14O6
E7500
meso-Erythritol, ≥99%
C4H10O4
73132
meso-Erythritol, analytical standard
C4H10O4
16355
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
HNO3
236241
Pentaerythritol, 99%
C5H12O4
P4755
Pentaerythritol, 98%
C5H12O4
430935
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
]n
182478
Poly(tetrafluoroethylene), powder, >40 μm particle size
]n
81377
Poly(tetrafluoroethylene), beads
]n