EMAIL THIS PAGE TO A FRIEND
PLoS Neglected Tropical Diseases
Minipool caprylic acid fractionation of plasma using disposable equipment: a practical method to enhance immunoglobulin supply in developing countries.
PMID 25719558