EMAIL THIS PAGE TO A FRIEND
Neurochemical research
C3 Polymorphism Influences Circulating Levels of C3, ASP and Lipids in Schizophrenic Patients.
PMID 25720829
Abstract
Excessive activation of complement is associated with many diseases including schizophrenia. Investigation of C3 polymorphisms, circulating C3, cleavage product ASP/C3adesArg, and lipid metabolism. Cross-sectional analysis. C3 genotyping (CC vs GG for R102L) was performed on 434 Tunisian people consisting of 272 schizophrenic (SZ) patients and 162 control subjects. In a age- and gender-matched subgroups of the three genotypes (131 SZ and 112 NOR), plasma triglycerides, total cholesterol (C), LDL-C, HDL-C, ASP, and complement C3 were measured. C3 gene polymorphism influences BMI and plasma C3, ASP, triglyceride, total cholesterol, LDL-C and HDL-C among SZ patients (p < 0.05-0.0001), with increasing values demonstrated from CC (common form) to CG (heterozygote form) to GG (rare homozygote) forms. Significant correlations between plasma C3 and BMI, triglyceride, HDL-C and ASP (p < 0.05-0.0001) were observed, while ASP correlated with BMI and LDL-C (p = 0.005, p = 0.001, respectively) in SZ patients. Further, proportional conversion of C3 to ASP (%ASP/C3) also increased (p < 0.0001, GG>CG>CC). C3 polymorphisms and plasma C3, ASP and %ASP/C3 correlated with lipid parameters in this SZ population, suggesting that factors predisposing patients to schizophrenia are permissive for complement pathway activation and dyslipidemic influences.