EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
Postsynaptic D2 dopamine receptor supersensitivity in the striatum of mice lacking TAAR1.
PMID 25721394
Abstract
Trace Amine-Associated Receptor 1 (TAAR1) is a G protein-coupled receptor (GPCR) known to modulate dopaminergic system through several mechanisms. Mice lacking this receptor show a higher sensitivity to dopaminergic stimuli, such as amphetamine; however, it is not clear whether D1 or D2 dopamine receptors and which associated intracellular signaling events are involved in this modulation. In the striatum of TAAR1 knock out (TAAR1-KO mice) we found that D2, but not D1, dopamine receptors were over-expressed, both in terms of mRNA and protein levels. Moreover, the D2 dopamine receptor-related G protein-independent AKT/GSK3 signaling pathway was selectively activated, as indicated by the decrease of phosphorylation of AKT and GSK3β. The decrease in phospho-AKT levels, suggesting an increase in D2 dopamine receptor activity in basal conditions, was associated with an increase of AKT/PP2A complex, as revealed by co-immunoprecipitation experiments. Finally, we found that the locomotor activation induced by the D2 dopamine receptor agonist quinpirole, but not by the full D1 dopamine receptor agonist SKF-82958, was increased in TAAR1-KO mice. These data demonstrate pronounced supersensitivity of postsynaptic D2 dopamine receptors in the striatum of TAAR1-KO mice and indicate that a close interaction of TAAR1 and D2 dopamine receptors at the level of postsynaptic structures has important functional consequences.