EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Gamisasangja-tang suppresses pruritus and atopic skin inflammation in the NC/Nga murine model of atopic dermatitis.
PMID 25721805
Abstract
Gamisasangja-tang (GST) is a traditional herbal formula prescribed for patients with intractable pruritus in association with various inflammatory skin diseases. To evaluate the effects of GST on pruritic skin inflammation and investigate its cellular and molecular mechanisms. We orally administered GST to NC/Nga (NC) mice, an animal model of atopic dermatitis. Scratching frequency and the dermatitis index were evaluated, and histological examination was performed using hematoxylin and eosin and toluidine blue staining. The levels of interleukin (IL)-31 and T-helper cell type 2 (TH2) cytokines were determined in both the dorsal skin and cultured splenocytes by real-time polymerase chain reaction (PCR) and enzyme-linked immunosorbent assay (ELISA), respectively. The serum levels of chemokines and immunoglobulin E (IgE) were determined by ELISA. Changes in the inflammatory cell population were analyzed by a hemocytometer. GST significantly lowered scratching frequency and inhibited increases in dermatitis index, thickness of epidermis/dermis and infiltration of chemokine (C-C motif) receptor 3 (CCR3)(+) and cluster of differentiation (CD)117(+)/FcεRIα (Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide)(+) cells in atopic skin. Both IL-31 mRNA expression and production were significantly reduced by GST, which was accomrease in the levels of IL-4, IL-5, and IL-13. Further, GST treatment suppressed the secretion of eotaxin, TARC (thymus and activation-regulated chemokine), IgE, and increases in the number of basophils and eosinophils in the blood. GST may have potential as an effective treatment for pruritic skin disease such as atopic dermatitis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
237329
1-Chloro-2,4-dinitrobenzene, ≥99%
C6H3ClN2O4
138630
1-Chloro-2,4-dinitrobenzene, 97%
C6H3ClN2O4