EMAIL THIS PAGE TO A FRIEND
International immunology
Dimethyl fumarate suppresses Theiler's murine encephalomyelitis virus-induced demyelinating disease by modifying the Nrf2-Keap1 pathway.
PMID 25721871
Abstract
Dimethyl fumarate (DMF) is a modifier of the nuclear factor (erythroid-derived 2)-2 (Nrf2)-kelch-like ECH-associated protein 1 (Keap1) pathway. DMF treatment in the effector phase significantly suppressed the development of Theiler's murine encephalomyelitis virus-induced demyelinating disease (TMEV-IDD) both clinically and histologically. DMF treatment leads to an enhanced Nrf2 antioxidant response in TMEV-IDD mice. DMF treatment in the effector phase significantly suppressed the level of IL-17A mRNA. DMF is known to inhibit differentiation of T helper 17 (Th17) cells via suppressing NF-κB. Taken together, our data suggest that DMF treatment in the effector phase may suppress TMEV-IDD not only via enhancing the antioxidant response but also via suppressing IL-17A.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU202811 MISSION® esiRNA, esiRNA targeting mouse Ptprc