EMAIL THIS PAGE TO A FRIEND
Food Chemistry
High performance liquid chromatography coupled to mass spectrometry for profiling and quantitative analysis of folate monoglutamates in tomato.
PMID 25722141