EMAIL THIS PAGE TO A FRIEND
Food chemistry
Effect of different aging techniques on the polysaccharide and phenolic composition and sensory characteristics of Syrah red wines fermented using different yeast strains.
PMID 25722146
Abstract
The effect of high levels of the polysaccharide Saccharomyces cerevisiae yeast strain (HPS) and another conventional yeast strain (FERM) on the polysaccharide and phenolic composition of Syrah red wines during alcoholic fermentation and subsequent aging on lees, with or without oak wood chips, and on inactive dry yeast was investigated. The HPS yeast released higher amounts of polysaccharides during alcoholic fermentation than FERM yeast (485 g L(-1) and 403 g L(-1), respectively) and after the aging period (516 g L(-1) and 500 g L(-1), respectively). The different aging techniques increased the polysaccharide concentration; the concentration was dependent on the aging technique applied. The interaction of the polysaccharides with the phenolic compounds depended on the yeast strain, aging technique, aging period and compound analysed. The HPS wines exhibited better sensory characteristics than the FERM wines after alcoholic fermentation; however, during the aging period, it was difficult to determine which technique produced the best wine due to the interactions of aging technique, aging period and sensory attribute evaluated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03910590
(±)-Catechin hydrate, primary pharmaceutical reference standard
C15H14O6 · xH2O
43412
(+)-Catechin, analytical standard
C15H14O6
79058
2-(4-Hydroxyphenyl)ethanol, analytical standard
C8H10O2
188255
2-(4-Hydroxyphenyl)ethanol, 98%
C8H10O2
T90301
3-(2-Hydroxyethyl)indole, 97%
C10H11NO
00070
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
C2H4O
402788
Acetaldehyde, ACS reagent, ≥99.5%
C2H4O
00071
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
C2H4O
W200336
Acetaldehyde, natural, ≥99%, FG
C2H4O
W200301
Acetaldehyde, ≥99%, meets FCC analytical specification
C2H4O
W200360
Acetaldehyde, ≥99%, meets FCC analytical specification
C2H4O
W200320
Acetaldehyde, ≥99%, FCC, stabilized
C2H4O
506788
Acetaldehyde, analytical standard
C2H4O
W200379
Acetaldehyde solution, 40 wt. % in H2O
C2H4O
W200387
Acetaldehyde solution, 40 wt. % in isopropanol
C2H4O
W200344
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
C2H4O
W200328
Acetaldehyde solution, 50 wt. % in ethanol
C2H4O
719099
Acetaldehyde solution, 5 M in THF
C2H4O
W200300
Acetaldehyde solution, natural, 50 wt. % in ethanol
C2H4O
74347
Hydroquinone, certified reference material, TraceCERT®
C6H6O2
1324002
Hydroquinone, United States Pharmacopeia (USP) Reference Standard
C6H6O2
PHR1469
Hydroquinone, Pharmaceutical Secondary Standard; Certified Reference Material
C6H6O2
72576
Myricetin, analytical standard
C15H10O8
M6760
Myricetin, ≥96.0%, crystalline
C15H10O8
70050
Myricetin, ≥96.0% (HPLC)
C15H10O8
36576
Sulfur, PESTANAL®, analytical standard
S