EMAIL THIS PAGE TO A FRIEND
Carbohydrate Research
Kinetic characterization of Aspergillus niger chitinase CfcI using a HPAEC-PAD method for native chitin oligosaccharides.
PMID 25723623
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F2252
L-(−)-Fucose, ≥99%
C6H12O5