EMAIL THIS PAGE TO A FRIEND
Biological Research
Fruits and barks extracts of Zanthozyllum heitzii a spice from Cameroon induce mitochondrial dependent apoptosis and Go/G1 phase arrest in human leukemia HL-60 cells.
PMID 25723957