EMAIL THIS PAGE TO A FRIEND
Febs Letters
C-type lectin-like domain and fibronectin-like type II domain of phospholipase A(2) receptor 1 modulate binding and migratory responses to collagen.
PMID 25724334