EMAIL THIS PAGE TO A FRIEND
Toxicology Letters
Effects of exposure to benzo[a]pyrene on metastasis of breast cancer are mediated through ROS-ERK-MMP9 axis signaling.
PMID 25724548
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1760
Benzo[a]pyrene, ≥96% (HPLC)
C20H12