EMAIL THIS PAGE TO A FRIEND
The international journal of biochemistry & cell biology
Early growth response-1 facilitates enterovirus 71 replication by direct binding to the viral genome RNA.
PMID 25724735
Abstract
Enterovirus 71 (EV71) infections can cause hand, foot and mouth disease (HFMD), meningoencephalitis, neonatal sepsis, and even fatal encephalitis in children. Unfortunately, there is currently no effective treatment for EV71 infection due to the lack of understanding of viral replication and infection; and viral infections have emerged as an imperative global hazard. Thus, it is extremely important to understand the mechanism of EV71 replication in order to prevent and control the diseases associated with EV71 infections. Early growth response-1 (EGR1) is a multifunctional transcription factor that regulates diverse biological functions, including inflammation, apoptosis, differentiation, tumorigenesis, and even viral infection. Here, we provide new insight into the role of EV71 infection in regulating EGR1 production; and reveal a novel mechanism by which EGR1 facilitates EV71 replication. We demonstrate that EV71 activates EGR1 expression during infection by stimulating the protein kinase A/protein kinase Cɛ/phosphoinositide 3-kinase/Akt (PKA/PKCɛ/PI3K/Akt) cascade. We further reveal that EV71-activated EGR1, in turn, regulates the internal ribosomal entry site (IRES) of EV71 to enhance viral replication. In addition, EGR1 facilitates EV71 replication by binding directly to stem-loops I and IV of EV71 5'-untranslated region (5'UTR) with its first two zinc fingers. Moreover, EGR1 protein co-localizes with EV71 RNA in the cytoplasm of infected cells to facilitate viral replication. Our results reveal an important new role of EGR1 in viral infection, provide new insight into the novel mechanism underlying the regulation of EV71 replication, and suggest a potential application of EGR1 in the control of EV71 infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU060821 MISSION® esiRNA, esiRNA targeting mouse Egr1 (esiRNA1)