EMAIL THIS PAGE TO A FRIEND
Theriogenology
Nuclei fluorescence microscopic observation on early embryonic development of mitogynogenetic diploid induced by hydrostatic pressure treatment in olive flounder (Paralichthys olivaceus).
PMID 25726149
Abstract
Sperm genetic material of olive flounder (Paralichthys olivaceus) was inactivated by ultraviolet irradiation. The nuclear phase changes during early embryonic development of diploid, haploid, and mitogynogenetic diploid induced by hydrostatic pressure treatment were observed under fluorescent microscope with 4',6-diamidino-2-phenylindole staining. The parameters of hydrostatic pressure treatment were 600 kg/cm(2) for 6 minutes at prometaphase stage. The data showed that developmental timing sequence of diploid and haploid fertilized eggs was similar. The cell cycle was about 48 minutes, including interphase (about 21 minutes), prophase (about 3 minutes), prometaphase (about 6 minutes), metaphase (about 6 minutes), anaphase (around 9 minutes), and telophase (about 3 minutes). After entering the fertilized egg, ultraviolet-inactivated sperm formed a male pronucleus and became a dense chromatin body in the cytoplasm. Dense chromatin body did not participate in nuclear division and unchanged all the time. For hydrostatic pressure-treated embryos, the first nuclear division and cytokinesis after treatment proceeded normally after about 15 minutes recovery. During the second mitosis, having undergone interphase, prophase, and prometaphase stage, chromosomes began to slowly spread around and scattered in the cell but not entered into metaphase and anaphase. The second nuclear division and cytokinesis was inhibited. The occurrence frequency of developmentally delayed embryos also showed that the second cleavage of about 80% treated eggs was inhibited. The inhibition of the second cleavage resulted to chromosome set doubling. So chromosome set doubling for mitogynogenetic flounder diploid induced by hydrostatic pressure treatment, performed at prometaphase stage, was mainly due to inhibition of the second mitosis rather than the first one.