EMAIL THIS PAGE TO A FRIEND
Mutation Research. Genetic Toxicology and Environmental Mutagenesis
Genotoxic and cytostatic effects of 6-pentadecyl salicylic anacardic acid in transformed cell lines and peripheral blood mononuclear cells.
PMID 25726174