EMAIL THIS PAGE TO A FRIEND
Biochemical and Biophysical Research Communications
Omega-3 free fatty acids inhibit tamoxifen-induced cell apoptosis.
PMID 25727020
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
08339
5-Aminolevulinic acid hydrochloride, ≥97.0% (AT)
C5H9NO3 · HCl