EMAIL THIS PAGE TO A FRIEND
Environmental Science & Technology
Effect of acid-catalyzed formation rates of benzimidazole-linked polymers on porosity and selective CO2 capture from gas mixtures.
PMID 25730399