EMAIL THIS PAGE TO A FRIEND
Pharmaceutical research
A comparison of the cholinergic activity of selected H2-antagonists and sulfoxide metabolites.
PMID 2573049
Abstract
Famotidine and selected H2-antagonists were evaluated with respect to toxicity and selected pharmacological activities. When administered intraperitoneally to mice at a dose equivalent to 10 times their respective H2-antagonist ED50 values, no deaths were observed. Similarly, no alteration in brain ACh concentrations or overt pharmacological effects were noted. However, at 400 mg/kg, ranitidine produced 89% lethality, followed by cimetidine (11%) and famotidine. Only cimetidine and famotidine at this dose significantly elevated brain acetylcholine levels. These results do not correlate with the in vitro data, where ORF-17578 and ranitidine were the most potent entities with respect to acetylcholinesterase inhibition (approximately 1-2 X 10(-6) M), followed by nizatidine greater than cimetidine greater than famotidine. The sulfoxide metabolites of ranitidine and cimetidine were approximately one-tenth as potent as their parent compounds with respect to inhibition of acetylcholinesterase. Direct muscarinic stimulation or potentiation of acetylcholine-induced contraction in ileal tissue was not observed for any of the H2-antagonists.