EMAIL THIS PAGE TO A FRIEND
General Physiology and Biophysics
The effect of long-term administered CRAC channels blocker on the functions of respiratory epithelium in guinea pig allergic asthma model.
PMID 25730896