EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Quantitative liquid chromatography-electrospray ionization-mass spectrometry analysis of amine-containing metabolites derivatized with cyanuric chloride and methylamine isotopologues.
PMID 25731825