EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
A robust smart window: reversibly switching from high transparency to angle-independent structural color display.
PMID 25732127
Abstract
A smart window is fabricated from a composite consisting of elastomeric poly(dimethylsiloxane) embedded with a thin layer of quasi-amorphous silica nanoparticles. The smart window can be switched from the initial highly transparent state to opaqueness and displays angle-independent structural color via mechanical stretching. The switchable optical property can be fully recovered after 1000 stretching/releasing cycles.