EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Polybutylene terephthalate-nickel oxide nanocomposite as a fiber coating.
PMID 25732309
Abstract
A highly efficient polybutylene terephthalate (PBT)-based nanocomposite containing nickel oxide nanoparticles was synthesized by electrospinning technique and used as a fiber coating for solid phase microextraction. The influential morphological parameters and capability of the prepared nanocomposite including the NiO content, the coating time, the PBT concentration and applied voltage were considered for optimization. The applicability of the synthesized fiber coating was examined by headspace solid phase micro extraction and gas chromatography mass spectrometry detection of some volatile organic compounds in aqueous samples. Among the synthesized nanocomposites and pristine PBT nanofibers, the fiber coating with 14% of NiO nanoparticles doping level exhibited the highest extraction efficiency. In addition, important parameters influencing the extraction/desorption process were investigated and optimized. The detection limits were less than 5 ng L(-1) using the selected ion monitoring mode. The inter-day and intra-day precisions of the developed method under optimized conditions were below 11%. The method showed a linearity in the range of 10-1000 ng L(-1) with the correlation coefficient greater than 0.9987. The optimized method was applied to the sampling of some volatile organic compounds from real water samples. The developed method is convenient, rapid, simple, easy and inexpensive and offers high sensitivity and sufficient reproducibility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03079
Ethylbenzene, analytical standard
C8H10
PHR1229
Ethylbenzene, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10
296848
Ethylbenzene, anhydrous, 99.8%
C8H10
E12508
Ethylbenzene, ReagentPlus®, 99%
C8H10
95670
m-Xylene, analytical standard
C8H10
95672
m-Xylene, puriss. p.a., ≥99.0% (GC)
C8H10
296325
m-Xylene, anhydrous, ≥99%
C8H10
185566
m-Xylene, ReagentPlus®, 99%
C8H10
PHR1313
m-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10
82762
Methanol, analytical standard
CH4O
PHR1372
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
CH4O
1424109
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
CH4O
95660
o-Xylene, analytical standard
C8H10
295884
o-Xylene, for HPLC, 98%
C8H10
X1040
o-Xylene, reagent grade, ≥98.0%
C8H10
PHR1312
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10
PHR1317
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
244511
Toluene, anhydrous, 99.8%
C7H8
648566
Toluene, for HPLC, ≥99.9%
C7H8
89680
Toluene, analytical standard
C7H8
24529
Toluene, puriss., ≥99.5% (GC)
C7H8
270377
Toluene, HPLC grade, 99.8%
C7H8
30-3305
Toluene, for HPLC
C7H8
30-3309
Toluene, JIS 5000, for residue analysis
C7H8
PHR1311
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8