EMAIL THIS PAGE TO A FRIEND
Analytica Chimica Acta
Multi-analyte high performance liquid chromatography coupled to high resolution tandem mass spectrometry method for control of pesticide residues, mycotoxins, and pyrrolizidine alkaloids.
PMID 25732310