EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
Quality evaluation and pattern recognition analyses of marker compounds from five medicinal drugs of Rutaceae family by HPLC/PDA.
PMID 25732613
Abstract
To establish a standard of quality control and to identify different origins for the Rutaceae family [Citri Unshiu Peel (CU), Citri Unshiu Immature Peel (CI), Ponciri Immature Fructus (PI), Aurantii Immature Fructus (AI), and Aurantii Fructus (AU)], 13 standards including rutin (1), narirutin (2), naringin (3), hesperidin (4), neohesperidin (5), neoponcirin (6), poncirin (7), naringenin (8), isosinensetin (9), sinensetin (10), nobiletin (11), heptamethoxyflavone (12), and tangeretin (13) were determined by high performance liquid chromatography (HPLC)/photo-diode array (PDA) analysis. A YMC ODS C18 (250 × 4.6 mm, 5 µm) column was used and the ratio of mobile phases of water (A) and acetonitrile (B) delivered to the column for gradient elution was applied. This method was fully validated with respect to linearity, accuracy, precision, stability, and robustness. The HPLC/PDA method was applied successfully to quantify 13 major compounds in the extracts of CU, CI, PI, AI, and AU. The pattern recognition analysis combined with LC chromatographic data was performed by repeated analysis of 27 reference samples in the above five Rutaceae oriental medicinal drugs. The established HPLC method was rapid and reliable for quantitative analysis and quality control of multiple components in five Rutaceae species with different origins.