EMAIL THIS PAGE TO A FRIEND
Analytica Chimica Acta
Highly selective and sensitive paper-based colorimetric sensor using thiosulfate catalytic etching of silver nanoplates for trace determination of copper ions.
PMID 25732695