EMAIL THIS PAGE TO A FRIEND
General and Comparative Endocrinology
The decapod red pigment-concentrating hormone (Panbo-RPCH) is the first identified neuropeptide of the order Plecoptera and is interpreted as homoplastic character state.
PMID 25733206