EMAIL THIS PAGE TO A FRIEND
Tumori
Plasma levels of E-cadherin and MMP-13 in prostate cancer patients: correlation with PSA, testosterone and pathological parameters.
PMID 25733385
Abstract
Prostate cancer is the most common malignant tumor in men. Serum prostate-specific antigen (PSA), Gleason score and clinical range at the time of diagnosis are important factors to predict prognosis and outcome after therapy but additional accurate and reliable biomarkers are still wanted. So far, few biomarkers of prostate cancer have been successfully implemented and are being used in clinical practice. However, modifications of E-cadherin and MMP-13 expression are likely to be involved in prostate cancer invasion and thus are potential biomarkers for prognosis. We analyzed the concentrations of E-cadherin and MMP-13 in plasma of patients with prostate cancer at diagnosis and 3 and 6 months after the beginning of treatment and related these measures to free and total PSA and other clinical features. The concentration of E-cadherin was lower in patients with prostate cancer compared to the control group, but there was no difference in the concentration of MMP-13 between these two groups. During treatment, however, we found no significant differences between the concentrations of MMP-13 and E-cadherin, but we observed a significant positive correlation between total PSA and E-cadherin plasma concentration at the third month of treatment and between total testosterone and MMP-13 plasma concentration before the start of treatment. The results suggest that these parameters could be used both in the diagnosis and prognosis of prostate cancer.