EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Proteome-wide analysis of mutant p53 targets in breast cancer identifies new levels of gain-of-function that influence PARP, PCNA, and MCM4.
PMID 25733866