EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Comprehensive analysis of heterotrimeric G-protein complex diversity and their interactions with GPCRs in solution.
PMID 25733868
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131377
1,10-Phenanthroline, ≥99%
C12H8N2
226947
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
C32H58N2O7S · xH2O
C6013
Cholesteryl hemisuccinate tris salt, anionic detergent
C31H50O4 · C4H11NO3
1295607
Glycerin, United States Pharmacopeia (USP) Reference Standard
C3H8O3
PHR1020
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O3
49779
Glycerol, tested according to Ph.Eur., anhydrous
C3H8O3
77067
Glycerol, analytical standard
C3H8O3
12146
Glycerol 3 M solution, 3 M
C3H8O3 · xH2O
63031
Magnesium ion solution for ISE, 0.01 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
63042
Magnesium ion solution for ISE, 0.1 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
71726
Sodium dodecyl sulfate, for ion pair chromatography, ≥99.0%
C12H25NaO4S
71727
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
C12H25NaO4S
Y0000620
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
C12H25NaO4S
71177
Sodium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClNa