EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Endocannabinoids are conserved inhibitors of the Hedgehog pathway.
PMID 25733905
Abstract
Hedgehog ligands control tissue development and homeostasis by alleviating repression of Smoothened, a seven-pass transmembrane protein. The Hedgehog receptor, Patched, is thought to regulate the availability of small lipophilic Smoothened repressors whose identity is unknown. Lipoproteins contain lipids required to repress Smoothened signaling in vivo. Here, using biochemical fractionation and lipid mass spectrometry, we identify these repressors as endocannabinoids. Endocannabinoids circulate in human and Drosophila lipoproteins and act directly on Smoothened at physiological concentrations to repress signaling in Drosophila and mammalian assays. Phytocannabinoids are also potent Smo inhibitors. These findings link organismal metabolism to local Hedgehog signaling and suggest previously unsuspected mechanisms for the physiological activities of cannabinoids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O0383
N-Oleoylethanolamine, ~98% (TLC)
C20H39NO2