EMAIL THIS PAGE TO A FRIEND
Nucleic Acid Therapeutics
siRNA delivery to the glomerular mesangium using polycationic cyclodextrin nanoparticles containing siRNA.
PMID 25734248
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56671
(±)-Propylene oxide, analytical standard
C3H6O