PLoS ONE
Serum-free and xenobiotic-free preservation of cultured human limbal epithelial cells.
PMID 25734654
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C106
CGS-12066 maleate salt, solid
C17H17F3N4 · C4H4O4
94563
Dimethyl sulfoxide, analytical standard
C2H6OS
01934
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
C2H6OS
1211006
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
C2H6OS
D2050000
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
C2H6OS
46043
Ethidium homodimer, suitable for fluorescence, ~90% (HPCE)
C46H50Cl4N8
54461
HEPES, ≥99.0% (T)
C8H18N2O4S
PHR1428
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
C8H18N2O4S
1316004
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
C21H30O5
PHR1014
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
C21H30O5
31719
Hydrocortisone, VETRANAL, analytical standard
C21H30O5
H1300000
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
C21H30O5
BP576
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
C21H30O5
Y0001092
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
C21H30O5
76523
L-Glutamine, certified reference material, TraceCERT®
C5H10N2O3
PHR1125
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
C5H10N2O3
209643
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Se
204307
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Se
229865
Selenium, powder, −100 mesh, 99.99% trace metals basis
Se
209651
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Se
HT-SE101
Selenium, pellets, < 5mm, ≥99.999%
Se
GF66395212
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Se