EMAIL THIS PAGE TO A FRIEND
Investigative Ophthalmology & Visual Science
The Role of Thrombin and Cell Contractility in Regulating Clustering and Collective Migration of Corneal Fibroblasts in Different ECM Environments.
PMID 25736789