EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Effects of RNA interference-mediated NRP-1 silencing on the proliferation and apoptosis of breast cancer cells.
PMID 25738638
Abstract
Lentiviral expression vectors carrying human NRP-1 short hairpin RNA (shRNA) were constructed and selected to present highly efficient NRP-1/shRNA interference sequences, in order to investigate the effects of RNA interference (RNAi)-mediated NRP-1 silencing on the biological activities of breast cancer cells. Three pairs of human NRP-1 targeted specific interference sequences and one pair of non-specific control sequences were designed, synthesized and subcloned into pLB lentiviral vectors, which were further identified by polymerase chain reaction (PCR) and sequencing. Recombinant and lentiviral packaging plasmids were co-transfected into 293FT cell lines in order to produce lentiviral particles and to infect breast cancer cells with high NRP-1 expression. Flow cytometry was used to sort green fluorescent protein-positive cells. Fluorescence quantitative-reverse transcription-PCR and western blot analysis were employed to identify the interference silencing sequence with the most efficient silencing profile. A cell counting kit-8 assay and an Annexin V-propidium iodide method in combination with flow cytometry were used to examine the effects of RNA interference-mediated NRP-1 gene silencing on cell proliferation, apoptosis and sensitivity to chemotherapy. The recombinant lentiviral plasmid pLB-NRP-1/shRNA was constructed successfully, as confirmed by PCR and sequencing. After the infection of recombinant lentiviral plasmids, the expression profiles of NRP-1 mRNA, and proteins of MCF-7 and SK-BR-3 cell-specific interference group (pLB-NRP-1/shRNA3) were significantly lower than that of the control group (P<0.05). Compared with the control group, the MCF-7 and SK-BR-3 cell-specific interference group (pLB-NRP-1/shRNA3) showed lower optical density values and higher apoptotic rates at 48, 72 and 96 h; these differences were statistically significant (P<0.05). EPI administration resulted in increased apoptosis in the MCF-7 and SK-BR-3 cell-specific interference groups compared with the control group (P<0.05). Lentiviral vectors encoding the human NRP-1 gene were constructed successfully and highly efficient NRP-1/shRNA interference sequences were selected. Furthermore, RNA interference (RNAi)-mediated NRP-1 silencing may induce proliferation suppression, apoptosis promotion, as well as enhanced sensitivity to chemotherapeutic agents.