EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
EGF Suppresses the Initiation and Drives the Reversion of TGF-β1-induced Transition in Hepatic Oval Cells Showing the Plasticity of Progenitor Cells.
PMID 25739869
Abstract
Transforming growth factor-β1 (TGF-β1) induces hepatic progenitors to tumor initiating cells through epithelial-mesenchymal transition (EMT), thus raising an important drawback for stem cell-based therapy. How to block and reverse TGF-β1-induced transition is crucial for progenitors' clinical application and carcinogenic prevention. Rat adult hepatic progenitors, hepatic oval cells, experienced E-cadherin to N-cadherin switch and changed to α-smooth muscle actin (α-SMA) positive cells after TGF-β1 incubation, indicating EMT. When TGF-β1 plus EGF were co-administrated to these cells, EGF dose-dependently suppressed the cadherin switch and α-SMA expression. Interestingly, if EGF was applied to TGF-β1-pretreated cells, the cells that have experienced EMT could return to their epithelial phenotype. Abruption of EGF receptor revealed that EGF exerted its blockage and reversal effects through phosphorylation of ERK1/2 and Akt. These findings suggest an important attribute of EGF on opposing and reversing TGF-β1 effects, indicating the plasticity of hepatic progenitors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4182
Tyrphostin AG 1478, ≥98%
C16H14ClN3O2