EMAIL THIS PAGE TO A FRIEND
Circulation journal : official journal of the Japanese Circulation Society
Secondary prevention strategy of cardiovascular disease using endothelial function testing.
PMID 25740088
Abstract
Over the past decades, secondary prevention of cardiovascular (CV) disease has improved and considerably reduced mortality rates. However, there remains a high-rate of new or recurrent CV events in those with established atherosclerotic vascular diseases. Although most of the prevailing therapies target the conventional risk factors, there is notable interindividual heterogeneity in adaptation to risk factors and response to therapies, which affects efficacy. It is desirable to have a methodology for directly assessing the functional significance of atherogenesis, and for managing individual patients based on their comprehensive vascular health. Endothelial function plays a pivotal role in all stages of atherosclerosis, from initiation to atherothrombotic complication. Endothelial function reflects the integrated effect of all the atherogenic and atheroprotective factors present in an individual, and is therefore regarded as an index of active disease process and a significant risk factor for future CV events. Moreover, improvement in endothelial function is associated with decreased risk of CV events, even in the secondary prevention setting. The introduction of endothelial function assessment into clinical practice may trigger the development of a more tailored and personalized medicine and improve patient outcomes. In this review, we summarize current knowledge on the contribution of endothelial dysfunction to atherosclerotic CV disease in the secondary prevention setting. Finally, we focus on the potential of an endothelial function-guided management strategy in secondary prevention.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1639
Patulin, ≥98.0% (HPLC)
C7H6O4
32759
Patulin, reference material
C7H6O4