EMAIL THIS PAGE TO A FRIEND
British journal of anaesthesia
High-sensitive cardiac troponin T measurements in prediction of non-cardiac complications after major abdominal surgery.
PMID 25740400
Abstract
Postoperative non-cardiac complication rates are as high as 11-28% after high-risk abdominal procedures. Emerging evidence indicates that postoperative cardiac troponinxa0T elevations are associated with adverse outcome in non-cardiac surgery. The aim of this study was to determine the relationship between postoperative high-sensitive cardiac troponin T elevations and non-cardiac complications in patients after major abdominal surgery. This prospective observational single-centre cohort study included patients at risk for coronary artery disease undergoing elective major abdominal surgery. Cardiac troponin was measured before surgery and at day 1, 3, and 7. Multivariable logistic regression analysis was performed to examine the adjusted association for different cut-off concentrations of postoperative myocardial injury and non-cardiac outcome. In 203 patients, 690 high-sensitive cardiac troponinxa0T measurements were performed. Fifty-three patients (26%) had a non-cardiac complication within 30 days after surgery. Hospital mortality was 4% (8/203). An increase in cardiac troponin T concentration ≥100% compared with baseline was a superior independent predictor of non-cardiac postoperative clinical complications (adjusted odds ratio 4.3, 95% confidence interval 1.8-10.1, P<0.001) and was associated with increased length of stay (9 days, 95% confidence interval 7-11 vs 7 days, 95% confidence interval 6-8, P=0.004) and increased hospital mortality (12 vs 2%, P=0.028). A postoperative high-sensitive cardiac troponin T increase ≥100% is a strong predictor of non-cardiac 30 day complications, increased hospital stay and hospital mortality in patients undergoing major abdominal surgery. NCT02150486.