EMAIL THIS PAGE TO A FRIEND
Life sciences
Multidrug resistance-associated protein 2 is involved in the efflux of Aconitum alkaloids determined by MRP2-MDCKII cells.
PMID 25744397
Abstract
Aconitum alkaloids mainly contain highly toxic aconitine (AC), mesaconitine (MA), and hypaconitine (HA) and less toxic benzoylaconine (BAC), benzoylmesaconine (BMA), benzoylhypaconine (BHA), aconine, mesaconine, and hypaconine. The efflux transporters including P-glycoprotein (P-gp), breast cancer resistance protein (BCRP) and multidrug resistance-associated protein 2 (MRP2) can efflux toxicants to prevent poisoning. Our previous publication has proved that P-gp and BCRP contributed to the efflux of AC, MA and HA, which is demonstrated in the human colonic adenocarcinoma cell lines (Caco-2 cells), Mardin-Darby canine kidney cell lines transfected with MDR1 or BCRP (MDR1-MDCKII and BCRP-MDCKII cells). However, the role of MRP2 remains uncertain. The MRP2-MDCKII cells were used to determine the efflux ratios (Er) and intracellular amounts of Aconitum alkaloids. In addition, the importance of MRP2 was further investigated with or without the MRP2 inhibitor, LTC4. The Er values of AC, MA, HA, BAC, BMA and BHA in MRP2-MDCKII cells (6.4 ± 0.3, 5.9 ± 0.5, 2.2 ± 0.2, 1.6 ± 0.3, 1.7 ± 0.2 and 1.9 ± 0.2 respectively) were significantly higher than those in MDCKII cells, which were close to 1. In the presence of LTC4, the Er values of AC, MA, HA, BAC, BMA and BHA were reduced to approximately 1 and their intracellular amounts were also significantly increased in MRP2-MDCKII cells. MRP2 was involved in the efflux of AC, MA, HA, BAC, BMA and BHA, which would be useful for the safe application of these components or their herbs.